Learn More
Cayman Chemical Guanosine 5'-diphosphate (sodium salt hydrate), Purity: ≥90%, 50 mg
SDP
Catalog No. 502992294
Encompass_Preferred
A purine nucleotide; precursor of GTP; has been used to study GTPases; activates the SUR2B/Kir6.1 channel in HEK293T cells in a patch-clamp assay at 100 µM
Catalog No. 50-299-2294 Supplier Cayman Chemical Supplier No. 3170450 MG
A purine nucleotide; precursor of GTP; has been used to study GTPases; activates the SUR2B/Kir6.1 channel in HEK293T cells in a patch-clamp assay at 100 µM