Learn More
Cayman Chemical GunosIn 5-monphosphates 5g
SDP
Supplier:  Cayman Chemical 169575 G
Encompass_Preferred
A nucleotide that upon phosphorylation to GTP becomes incorporated into RNA; used to study modulation of glutamatergic neurotransmission
Catalog No. 50-296-4949