Learn More
Cayman Chemical GunosIn 5-trIphosphateso 50mg
SDP
Catalog No. 502973119
Encompass_Preferred
A substrate for the synthesis of RNA during the transcription process and the synthesis of DNA during DNA replication; participates in G-protein-related signal transduction where it is converted to GDP through the action of GTPases
Catalog No. 50-297-3119 Supplier Cayman Chemical Supplier No. 1606050 MG
A substrate for the synthesis of RNA during the transcription process and the synthesis of DNA during DNA replication; participates in G-protein-related signal transduction where it is converted to GDP through the action of GTPases