Learn More
Cayman Chemical Guanosine 5'-[-thio]triphosphate (lithium salt) 10mg
SDP
Catalog No. 502911527
Encompass_Preferred
A hydrolysis-resistant analog of GTP; binds to and activates G proteins in the presence of magnesium; has been used to obtain crystal structures of GTPases in their active states, as well as measure the stability of GTPases in the presence of ligands; [35S]GTPγS is commonly used in binding assays to determine agonism or antagonism at GPCRs
Catalog No. 50-291-1527 Supplier Cayman Chemical Supplier No. 3509810 MG
A hydrolysis-resistant analog of GTP; binds to and activates G proteins in the presence of magnesium; has been used to obtain crystal structures of GTPases in their active states, as well as measure the stability of GTPases in the presence of ligands; [35S]GTPγS is commonly used in binding assays to determine agonism or antagonism at GPCRs