Learn More
Cayman Chemical GunosIn 5thIotrIphosp 2.7mg
SDP
Catalog No. 502918998
Encompass_Preferred
A hydrolysis-resistant analog of GTP; binds to and activates G proteins in the presence of magnesium; has been used to obtain crystal structures of GTPases in their active states, as well as measure the stability of GTPases in the presence of ligands; [35S]GTPγS is commonly used in binding assays to determine agonism or antagonism at GPCRs
Catalog No. 50-291-8998 Supplier Cayman Chemical Supplier No. 385652.7 MG
A hydrolysis-resistant analog of GTP; binds to and activates G proteins in the presence of magnesium; has been used to obtain crystal structures of GTPases in their active states, as well as measure the stability of GTPases in the presence of ligands; [35S]GTPγS is commonly used in binding assays to determine agonism or antagonism at GPCRs