Learn More
Cayman Chemical GunosIn 5trIphosphATso 100mg
SDP
Catalog No. 502968604
Encompass_Preferred
A substrate for the synthesis of RNA during the transcription process and the synthesis of DNA during DNA replication; participates in G-protein-related signal transduction where it is converted to GDP through the action of GTPases
Catalog No. 50-296-8604 Supplier Cayman Chemical Supplier No. 16060100 MG
A substrate for the synthesis of RNA during the transcription process and the synthesis of DNA during DNA replication; participates in G-protein-related signal transduction where it is converted to GDP through the action of GTPases