Learn More
Cayman Chemical Guanosine-5'-O-triphosphoro-γ-1-(5-sulfonic acid)naphthylamidate (sodium salt), Purity: ≥95%, 3.6 mg
SDP
Catalog No. 502919086
Encompass_Preferred
A fluorescent derivative of GTP; has been used as a substrate for snake venom PDEs and to monitor tubulin assembly in cell-free assays; ex/em = 340/460 nm, respectively
Catalog No. 50-291-9086 Supplier Cayman Chemical Supplier No. 386873.6 MG
A fluorescent derivative of GTP; has been used as a substrate for snake venom PDEs and to monitor tubulin assembly in cell-free assays; ex/em = 340/460 nm, respectively