Learn More
Electron Microscopy Sciences Forensic Mounts Gunshot Residue Lab Kit, 8 mm
SDP
Supplier:  Electron Microscopy Sciences 7648018
Encompass_Preferred
Includes ten 12.7 mm SEM pin mounts in clean, glass storage tubes (25 mm OD x 55 mm high) secured in a compact transport box with labels. This Kit is designed to avoid any contamination of GSR samples. Choose from either standard 12.7 mm pin mounts with 8 mm pins or with shorter 6 mm pins. Also available as a sample kit without mounts and tabs.
Pin length: 8 mm
Catalog No. 50-192-7160