Learn More
Electron Microscopy Sciences Forensic Mounts Gunshot Residue Lab Kit ZEISS/LEO, 6 mm
SDP
Catalog No. 501927161
Encompass_Preferred
Includes ten 12.7 mm SEM pin mounts in clean, glass storage tubes (25 mm OD x 55 mm high) secured in a compact transport box with labels. This Kit is designed to avoid any contamination of GSR samples. Choose from either standard 12.7 mm pin mounts with 8 mm pins or with shorter 6 mm pins. Also available as a sample kit without mounts and tabs.
Pin length: 6 mm
Catalog No. 50-192-7161 Supplier Electron Microscopy Sciences Supplier No. 7648019
Includes ten 12.7 mm SEM pin mounts in clean, glass storage tubes (25 mm OD x 55 mm high) secured in a compact transport box with labels. This Kit is designed to avoid any contamination of GSR samples. Choose from either standard 12.7 mm pin mounts with 8 mm pins or with shorter 6 mm pins. Also available as a sample kit without mounts and tabs.
Pin length: 6 mm