Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Gunshot Residue Presumptive Test Kit
GREENER_CHOICESCI_ED

Catalog No. s96637 Shop All Aldon Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96637 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid and Lead and Lead Compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S96637 Supplier Innovating Science™ Supplier No. IS9001
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid and Lead and Lead Compounds, which are known to the State of California to cause cancer and reproductive harm.

Includes

Instructions, diphenylamine/sulfuric acid solution UN1830 (5mL), sodium rhodizonate (0.025 g), lead nitrate (0.05M aqueous solution, 5mL), 20 alcohol swabs. Enough materials for 30 tests.

Determine if a surface has been exposed to a discharged firearm.

Two-part test with a rapid color change to verify the presence of nitrates and lead.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) For presumptive test
Includes Instructions, 5mL diphenylamine sulfuric acid solution un1830, 0.025 g sodium rhodizonate, 5mL lead nitrate, aqueous solution 0.05m, 20 alcohol swabs, teacher manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 30 Students
Product Type Gun Shot Residue Presumptive Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.