Learn More
Cayman Chemical Guanethidine (sulfate), Purity: ≥95%, CAS: 645-43-2, 100 mg
SDP
Catalog No. 502970748
Encompass_Preferred
A peripherally acting, antihypertensive compound that competes with norepinephrine for transport into presynaptic terminals of adrenergic neurons by the norepinephrine transporter, reducing the release of norepinephrine in response to action potentials
Catalog No. 50-297-0748 Supplier Cayman Chemical Supplier No. 16217100 MG
A peripherally acting, antihypertensive compound that competes with norepinephrine for transport into presynaptic terminals of adrenergic neurons by the norepinephrine transporter, reducing the release of norepinephrine in response to action potentials