Learn More
Cayman Chemical Guanylate Cyclase β1 subunit (soluble) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502968791
Encompass_Preferred
Immunogen: Synthetic peptide from an internal region of rat sGC β1 subunit • Host: Rabbit • Species Reactivity: (+) Human, bovine, rat • Cross Reactivity: (–) sGC α1 subunit • Applications: IHC, WB
Catalog No. 50-296-8791 Supplier Cayman Chemical Supplier No. 1608971 EA
Immunogen: Synthetic peptide from an internal region of rat sGC β1 subunit • Host: Rabbit • Species Reactivity: (+) Human, bovine, rat • Cross Reactivity: (–) sGC α1 subunit • Applications: IHC, WB