Learn More
Cayman Chemical Gunylate Cyclase 1 subunIt 1ea
SDP
Catalog No. 502912803
Encompass_Preferred
A clickable form of uridine is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo
Catalog No. 50-291-2803 Supplier Cayman Chemical Supplier No. 3608971 EA
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
A clickable form of uridine is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo