Learn More
Cayman Chemical Guanylate Cyclase α subunit (soluble) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502971219
Encompass_Preferred
Immunogen: Peptide affinity-purified antibody • Host: Rabbit • Species Reactivity: (+) Human, bovine, and mouse • Applications: WB and IHC
Catalog No. 50-297-1219 Supplier Cayman Chemical Supplier No. 1608951 EA
Immunogen: Peptide affinity-purified antibody • Host: Rabbit • Species Reactivity: (+) Human, bovine, and mouse • Applications: WB and IHC