Learn More
Cayman Chemical Gunylate Cyclase a subunIt 1ea
SDP
Catalog No. 502909193
Encompass_Preferred
A fluorogenic substrate for DPP-4 AMC is released upon enzymatic cleavage by DPP-4 and its fluorescence can be used to quantify DPP-4 activity ex/em 340-360/440-460 nm respectively
Catalog No. 50-290-9193 Supplier Cayman Chemical Supplier No. 3608951 EA
A fluorogenic substrate for DPP-4 AMC is released upon enzymatic cleavage by DPP-4 and its fluorescence can be used to quantify DPP-4 activity ex/em 340-360/440-460 nm respectively