Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guretolimod hydrochloride | 00-00-0 | 98.7% | 550.01 | C24H35ClF3N5O4 | 5 MG
SDP

Catalog No. 5000222218
Encompass_Preferred

Guretolimod hydrochloride is a synthetic small-molecule Toll-like receptor 7 (TLR7) agonist supplied for research use. It is used in preclinical studies to stimulate innate immune signaling and evaluate immunostimulatory and antitumor responses in vitro and in vivo models.

  • Potent TLR7 agonist for immune activation studies.
  • Suitable for in vitro and in vivo preclinical research.
  • High purity (approximately 98.7%) for consistent results.
  • Hydrochloride salt form to improve solubility and handling.
  • Characterized chemical identity with molecular formula C24H35ClF3N5O4 and molecular weight 550.01.

Catalog No. 50-002-22218 Supplier Medchemexpress LLC Supplier No. HY139575A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guretolimod hydrochloride is a synthetic small-molecule Toll-like receptor 7 (TLR7) agonist supplied for research use. It is used in preclinical studies to stimulate innate immune signaling and evaluate immunostimulatory and antitumor responses in vitro and in vivo models.

  • Potent TLR7 agonist for immune activation studies.
  • Suitable for in vitro and in vivo preclinical research.
  • High purity (approximately 98.7%) for consistent results.
  • Hydrochloride salt form to improve solubility and handling.
  • Characterized chemical identity with molecular formula C24H35ClF3N5O4 and molecular weight 550.01.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.