Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guretolimod hydrochloride | 00-00-0 | 98.7% | C24H35ClF3N5O4 | 10MG
SDP

Catalog No. 5000260574
Encompass_Preferred

Guretolimod hydrochloride is a Toll-like receptor 7 (TLR7) agonist supplied as the hydrochloride salt for research use. It is intended for laboratory studies probing TLR7-mediated immune activation and innate immune signaling; not for human or clinical use.

  • High chemical purity (98.67%).
  • Molecular formula C24H35ClF3N5O4 with molecular weight 550.01 Da.
  • Selective TLR7 agonist activity for immune-response research.
  • Provided as a hydrochloride salt to aid solubility in common solvents.
  • Available in small milligram pack sizes suitable for screening.

Catalog No. 50-002-60574 Supplier Medchemexpress LLC Supplier No. HY139575A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guretolimod hydrochloride is a Toll-like receptor 7 (TLR7) agonist supplied as the hydrochloride salt for research use. It is intended for laboratory studies probing TLR7-mediated immune activation and innate immune signaling; not for human or clinical use.

  • High chemical purity (98.67%).
  • Molecular formula C24H35ClF3N5O4 with molecular weight 550.01 Da.
  • Selective TLR7 agonist activity for immune-response research.
  • Provided as a hydrochloride salt to aid solubility in common solvents.
  • Available in small milligram pack sizes suitable for screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.