Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guretolimod (hydrochloride) | 98.7% | 550.01 | 25 MG
SDP

Catalog No. 5000349564
Encompass_Preferred

Guretolimod hydrochloride is a Toll-like receptor 7 (TLR7) agonist. It is used in laboratory settings for research purposes, particularly in the study of immunology and inflammation.

  • Toll-like receptor 7 (TLR7) agonist
  • Molecular weight of 550.01
  • Molecular formula C24H35ClF3N5O4
  • Off-white to light yellow solid appearance
  • Purity of 98.7% by HPLC
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at -20°C in sealed container, away from moisture and light

Catalog No. 50-003-49564 Supplier Medchemexpress LLC Supplier No. 25MGHY139575A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guretolimod hydrochloride is a Toll-like receptor 7 (TLR7) agonist. It is used in laboratory settings for research purposes, particularly in the study of immunology and inflammation.

  • Toll-like receptor 7 (TLR7) agonist
  • Molecular weight of 550.01
  • Molecular formula C24H35ClF3N5O4
  • Off-white to light yellow solid appearance
  • Purity of 98.7% by HPLC
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at -20°C in sealed container, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.