Learn More
Gurley Field Finder
SDP
Catalog No. 12454 Shop All Gurley Precision Instruments Products
Click to view available options
Dimensions (L x W):
3 x 1 in.
Catalog No. Dimensions (L x W)
12-454 3 x 1 in.
1 options
Catalog No. 12-454 Supplier Gurley Supplier No. 7100
Microscopic road map
  • Each square is marked with letter and number
  • Center detail in field of microscope, replace specimen slide with field finder, and read coordinates
Specifications
Dimensions (L x W) 3 x 1 in.
For Use With (Application) Relocate fields of interest in a slide-mounted specimen