Learn More
Cayman Chemical Gusacitinib, Purity: ≥98%, CAS: 1425381-60-7, 10 mg
SDP
Catalog No. 502913916
Encompass_Preferred
A dual inhibitor of JAKs (IC50s = 46, 4, and 11 nM for JAK1-3, respectively) and Syk family kinases (IC50s = 5 and 8 nM for SyK and TYK2, respectively)
Catalog No. 50-291-3916 Supplier Cayman Chemical Supplier No. 3655410 MG
A dual inhibitor of JAKs (IC50s = 46, 4, and 11 nM for JAK1-3, respectively) and Syk family kinases (IC50s = 5 and 8 nM for SyK and TYK2, respectively)