Learn More
Aviva Systems Biology Corp GUSB Antibody - C-terminal region (ARP44234_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44234_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GUSB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GUSB. Alternate names: MPS7, BGApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58753