Learn More
Medchemexpress LLC Guselkumab 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP99315MG
Encompass_Preferred
Guselkumab is a recombinant human IgG1 monoclonal antibody against the IL-23p19 subunit Guselkumab binds to human and cynomolgus monkey IL-23 with Kd values of 3 3 and 1 9 pmol/L respectively Guselkumab inhibits production of cytokines lying downstream of the IL-23 signaling pathway and can be used for psoriatic arthritis research[1] /br
Catalog No. 50-003-23506