Learn More
Ted Pella Inc GUST Spray Anyway 360° Duster, 8 oz.
SDP
Catalog No. NC0866659
Encompass
GUST 360° Spray Anyway Duster lets you spray from any angle - even upside down- without frosting. Contains Dustex 11, a 100% ozone safe, non-abrasive, inert, moisture-free, ultra clean gas for precision cleaning, no CFC's, no HCFC's. Suitable for removing dust, lint and particulates from microscopes, cameras, precision parts, electronic equipment and other delicate parts from any angle. Delivers up to 80 psi blasts. 8 oz. (187ml) canister.
Catalog No. NC0866659 Supplier Ted Pella Inc Supplier No. 81410
GUST 360° Spray Anyway Duster lets you spray from any angle - even upside down- without frosting. Contains Dustex 11, a 100% ozone safe, non-abrasive, inert, moisture-free, ultra clean gas for precision cleaning, no CFC's, no HCFC's. Suitable for removing dust, lint and particulates from microscopes, cameras, precision parts, electronic equipment and other delicate parts from any angle. Delivers up to 80 psi blasts. 8 oz. (187ml) canister.