Learn More
STONER INC GUST EZ DSTR 12/12OZ AROSL CS
Catalog No. NC9452572
Encompass_Preferred
Contains 12 oz. of 100% pure difluoroethane. Blasting power exceeds 70 psi. Non-ozone depleting, and filtered to 0.2 microns.
Catalog No. NC9452572 Supplier STONER INC Supplier No. 94203CSTON01
Contains 12 oz. of 100% pure difluoroethane. Blasting power exceeds 70 psi. Non-ozone depleting, and filtered to 0.2 microns.