Learn More
STONER INC GUST PREMIUM AIR DUSTR 12/15OZ
Catalog No. NC9634330
Encompass_Preferred
Contains 15 oz. of 100% pure tetrafluoroethane. Blasting power exceeds 80 psi. Non-flammable, non-ozone depleting, and filtered to 0.2 microns.
This item is not returnable. View return policy
Catalog No. NC9634330 Supplier STONER INC Supplier No. 94153CSTON01
Contains 15 oz. of 100% pure tetrafluoroethane. Blasting power exceeds 80 psi. Non-flammable, non-ozone depleting, and filtered to 0.2 microns.