Learn More
Analytical Sales & Services Gusto High-Speed Mini-Centrifuge
Catalog No. 5000804842
Encompass
Gusto High-Speed Mini-Centrifuge\n• 110-240 VAC, 50/60 H\n• Speed: 500-12,500 rpm\n• RFC: 15-9, 800 Xg (203 x 171 x 114 mm)\n\nQty/Pkg: EACH
Catalog No. 50-008-04842 Supplier Analytical Sales & Services Supplier No. 6100506
Gusto High-Speed Mini-Centrifuge\n• 110-240 VAC, 50/60 H\n• Speed: 500-12,500 rpm\n• RFC: 15-9, 800 Xg (203 x 171 x 114 mm)\n\nQty/Pkg: EACH