Learn More
ATCC Gut Microbiome; Genomic Mix
Catalog No. 502381974
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises an even mixture of genomic DNA prepared from fully sequenced, characterized, and authenticated bacterial species observed in normal and atypical gut microbial communities. Each order for this product is provided with access to run 20 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 2.4x108 genome copies/vial ± 1 log* *Genomic DNA concentrations indicate ATCC manufacturing specifications and are provided as a reference only
Catalog No. 50-238-1974 Supplier ATCC Supplier No. MSA1006
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises an even mixture of genomic DNA prepared from fully sequenced, characterized, and authenticated bacterial species observed in normal and atypical gut microbial communities. Each order for this product is provided with access to run 20 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 2.4x108 genome copies/vial ± 1 log* *Genomic DNA concentrations indicate ATCC manufacturing specifications and are provided as a reference only