Learn More
Medchemexpress LLC Gut restricted-7 | 2553218-46-3 | 98.6% | C25H40FNaO6S | 10MG
SDP
Catalog No. 5000258946
Encompass_Preferred
Gut restricted-7 (GR-7) is a potent, covalent, orally active pan-bile salt hydrolase (BSH) inhibitor that is tissue-selective and largely restricted to the gut. It decreases gut bacterial BSH activity and lowers deconjugated bile acid levels in mouse feces, and is provided for laboratory research use only.
  • Potent covalent pan-BSH inhibitor.
  • Orally active and tissue-selective for the gut.
  • Reduces deconjugated bile acid levels in feces.
  • High chemical purity (≈98.6%).
  • Available as solid and as 10 mM solution in DMSO.
  • Store solid sealed at 4°C; in solution store at -80°C or -20°C.
Catalog No. 50-002-58946 Supplier Medchemexpress LLC Supplier No. HY13574710MG
Gut restricted-7 (GR-7) is a potent, covalent, orally active pan-bile salt hydrolase (BSH) inhibitor that is tissue-selective and largely restricted to the gut. It decreases gut bacterial BSH activity and lowers deconjugated bile acid levels in mouse feces, and is provided for laboratory research use only.
  • Potent covalent pan-BSH inhibitor.
  • Orally active and tissue-selective for the gut.
  • Reduces deconjugated bile acid levels in feces.
  • High chemical purity (≈98.6%).
  • Available as solid and as 10 mM solution in DMSO.
  • Store solid sealed at 4°C; in solution store at -80°C or -20°C.