Learn More
Medchemexpress LLC Gut restricted-7 (GR-7) | 2553218-46-3 | MFCD00009109 | 98.6% | 510.63 | C25H40FNaO6S | 5 MG
SDP
Catalog No. 5000222102
Encompass_Preferred
Gut restricted-7 (GR-7) is a potent, covalent, orally active pan-bile salt hydrolase (BSH) inhibitor designed for gut-restricted activity in research settings. It decreases gut bacterial BSH activity and reduces levels of deconjugated bile acids in feces in animal studies. Supplied as a high-purity solid, it is intended for in vitro and in vivo experiments.
Catalog No. 50-002-22102 Supplier Medchemexpress LLC Supplier No. HY1357475MG
Gut restricted-7 (GR-7) is a potent, covalent, orally active pan-bile salt hydrolase (BSH) inhibitor designed for gut-restricted activity in research settings. It decreases gut bacterial BSH activity and reduces levels of deconjugated bile acids in feces in animal studies. Supplied as a high-purity solid, it is intended for in vitro and in vivo experiments.