Learn More
Med Vet International Ethicon Surgical Gut Suture - Plain, 5-0, Yellowish Tan, 1X18" G-3 Double Armed, Absorbable, 13mm, 12/box
Catalog No. 503045008
Encompass_Preferred
The Ethicon 772G suture is a high-quality absorbable plain gut surgical suture. It measures 18" in length, has a size of 5-0, and are dyed yellowish tan to provide greater visibility while suturing. This suture comes with a G-3 double armed reverse cutting edge needle and has 12 sutures in a box.
Catalog No. 50-304-5008 Supplier Med Vet International Supplier No. 772G
The Ethicon 772G suture is a high-quality absorbable plain gut surgical suture. It measures 18" in length, has a size of 5-0, and are dyed yellowish tan to provide greater visibility while suturing. This suture comes with a G-3 double armed reverse cutting edge needle and has 12 sutures in a box.