Learn More
Sklar Instruments Guthrie Double Hook, Sklar 5 in., 2 Long Prongs Sharp, Stainless Steel. EA.
SDP
Catalog No. 501184112 Shop All Sklar Instruments Products
Encompass_Preferred
Guthrie Double Hook, Sklar. 5 in., 2 Long Prongs Sharp, Stainless Steel. Merit Guthrie Hook is a retractor used in delicate procedures like plastic surgery to reduce trauma. The two prongs are delicate and sharp with a 1.5mm width between them. The overall length is 5 inches.
Catalog No. 50-118-4112 Supplier Sklar Instruments Supplier No. 98821
Guthrie Double Hook, Sklar. 5 in., 2 Long Prongs Sharp, Stainless Steel. Merit Guthrie Hook is a retractor used in delicate procedures like plastic surgery to reduce trauma. The two prongs are delicate and sharp with a 1.5mm width between them. The overall length is 5 inches.