Learn More
Medchemexpress LLC Guvacine hydrobromide | 6027-92-5 | 5 MG
SDP
Catalog No. 5000421345
Encompass_Preferred
Guvacine hydrobromide is an alkaloid naturally found in the nut of Areca catechu and functions as a potent GABA uptake inhibitor. It is a solid with a white to off-white appearance and boasts a high purity of 99.86%. This product is strictly for research use.
  • Potent GABA uptake inhibitor
  • Inhibits rat GAT-1, rat GAT-2, and rat GAT-3 with specific IC50 values
  • Inhibits human GAT-1, human GAT-3, and human BGT-3 with specific IC50 values
  • 99.86% purity
  • Solid, white to off-white appearance
  • Intended for research use only
Catalog No. 50-004-21345 Supplier Medchemexpress LLC Supplier No. HYN2482A5MG
Guvacine hydrobromide is an alkaloid naturally found in the nut of Areca catechu and functions as a potent GABA uptake inhibitor. It is a solid with a white to off-white appearance and boasts a high purity of 99.86%. This product is strictly for research use.
  • Potent GABA uptake inhibitor
  • Inhibits rat GAT-1, rat GAT-2, and rat GAT-3 with specific IC50 values
  • Inhibits human GAT-1, human GAT-3, and human BGT-3 with specific IC50 values
  • 99.86% purity
  • Solid, white to off-white appearance
  • Intended for research use only