Learn More
Medchemexpress LLC Guvacine hydrochloride | 6027-91-4 | 99.9% | 163.60 g·mol⁻¹ | C6H10ClNO2 | 100 MG
SDP
Catalog No. 5000211166
Encompass_Preferred
Guvacine hydrochloride is a research reagent used to inhibit GABA uptake in neuropharmacology and biochemical studies. It is supplied as a white to off-white solid and is available in both solid and small-solution formats for in vitro experiments.
  • Functions as a GABA uptake inhibitor for neuropharmacology research.
  • High purity (~99.9%) minimizes impurities in assays.
  • Available as solids and as a DMSO solution for experimental flexibility.
  • White to off-white solid suitable for analytical and in vitro use.
  • Molecular weight 163.60 g·mol⁻¹ supports accurate stoichiometry calculations.
Catalog No. 50-002-11166 Supplier Medchemexpress LLC Supplier No. HY100809100MG
Guvacine hydrochloride is a research reagent used to inhibit GABA uptake in neuropharmacology and biochemical studies. It is supplied as a white to off-white solid and is available in both solid and small-solution formats for in vitro experiments.
  • Functions as a GABA uptake inhibitor for neuropharmacology research.
  • High purity (~99.9%) minimizes impurities in assays.
  • Available as solids and as a DMSO solution for experimental flexibility.
  • White to off-white solid suitable for analytical and in vitro use.
  • Molecular weight 163.60 g·mol⁻¹ supports accurate stoichiometry calculations.