Learn More
Medchemexpress LLC Guvacine hydrochloride | 6027-91-4 | MFCD00055191 | 99.9% | 163.60 g/mol | C6H10ClNO2 | 1 ML
SDP
Catalog No. 5000213271
Encompass_Preferred
Guvacine hydrochloride is a GABA transporter inhibitor used in neuroscience research to investigate GABAergic signaling and transporter pharmacology. It is supplied with documented purity, molecular data, and recommended storage conditions to support reliable in vitro experiments.
  • Inhibits GABA transporter activity with reported IC50 values for multiple transporter subtypes.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 ML) for immediate use in assays.
  • Also available as a solid for custom preparation and long-term storage.
  • High reported purity supports reproducible experimental results.
  • Storage guidance provided to maintain stability of solid and solution forms.
Catalog No. 50-002-13271 Supplier Medchemexpress LLC Supplier No. HY10080910MM 1ML
Guvacine hydrochloride is a GABA transporter inhibitor used in neuroscience research to investigate GABAergic signaling and transporter pharmacology. It is supplied with documented purity, molecular data, and recommended storage conditions to support reliable in vitro experiments.
  • Inhibits GABA transporter activity with reported IC50 values for multiple transporter subtypes.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 ML) for immediate use in assays.
  • Also available as a solid for custom preparation and long-term storage.
  • High reported purity supports reproducible experimental results.
  • Storage guidance provided to maintain stability of solid and solution forms.