Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guvacine hydrochloride | 6027-91-4 | MFCD00055191 | 99.9% | 163.60 g/mol | C6H10ClNO2 | 25 MG
SDP

Catalog No. 5000212285
Encompass_Preferred

Guvacine hydrochloride is the hydrochloride salt of guvacine, a pyridine alkaloid that acts as a GABA uptake inhibitor. Supplied as a high-purity research reagent, it is used in biochemical and neuropharmacological studies to probe GABA transporter activity. The substance is intended for laboratory research use only and is not for human or veterinary use.

  • High purity (99.93%) for reliable experimental results.
  • Molecular weight 163.60 g/mol.
  • Molecular formula C6H10ClNO2.
  • CAS number 6027-91-4 for unambiguous identification.
  • Available in multiple pack sizes and solution formats for flexible use.
  • Suitable for in vitro biochemical and pharmacology research.

Catalog No. 50-002-12285 Supplier Medchemexpress LLC Supplier No. HY10080925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guvacine hydrochloride is the hydrochloride salt of guvacine, a pyridine alkaloid that acts as a GABA uptake inhibitor. Supplied as a high-purity research reagent, it is used in biochemical and neuropharmacological studies to probe GABA transporter activity. The substance is intended for laboratory research use only and is not for human or veterinary use.

  • High purity (99.93%) for reliable experimental results.
  • Molecular weight 163.60 g/mol.
  • Molecular formula C6H10ClNO2.
  • CAS number 6027-91-4 for unambiguous identification.
  • Available in multiple pack sizes and solution formats for flexible use.
  • Suitable for in vitro biochemical and pharmacology research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.