Learn More
Cayman Chemical Guvacine (hydrochloride), Purity: ≥95%, CAS: 6027-91-4, 100 mg
SDP
Catalog No. 502993754
Encompass_Preferred
An inhibitor of GABA uptake (IC50 = 10 μM; Ki = 14 μM) in rat hippocampal brain slices; decreases spontaneous activity in mice; decreases tail flick reaction time in a rat model of morphine analgesia
Catalog No. 50-299-3754 Supplier Cayman Chemical Supplier No. 23361100 MG
An inhibitor of GABA uptake (IC50 = 10 μM; Ki = 14 μM) in rat hippocampal brain slices; decreases spontaneous activity in mice; decreases tail flick reaction time in a rat model of morphine analgesia