Learn More
Medchemexpress LLC Guvacoline hydrobrom 10mg | 17210-51-4 | 222.08 g·mol⁻1 | C7H12BrNO2 | 10 MG
SDP
Catalog No. 5000308223
Encompass_Preferred
Guvacoline hydrobromide is the hydrobromide salt of guvacoline, a pyridine alkaloid that acts as a weak full agonist at atrial and ileal muscarinic acetylcholine receptors. This research-grade solid is provided for in vitro pharmacology and receptor-binding studies.
  • High purity (96.09%) research-grade material.
  • Molecular weight 222.08 g·mol⁻1; formula C7H12BrNO2.
  • Suitable for muscarinic receptor pharmacology and neuropharmacology assays.
  • Supplied as a 10 mg solid for small-scale experimental use.
Catalog No. 50-003-08223 Supplier Medchemexpress LLC Supplier No. HY11649010MG
Guvacoline hydrobromide is the hydrobromide salt of guvacoline, a pyridine alkaloid that acts as a weak full agonist at atrial and ileal muscarinic acetylcholine receptors. This research-grade solid is provided for in vitro pharmacology and receptor-binding studies.
  • High purity (96.09%) research-grade material.
  • Molecular weight 222.08 g·mol⁻1; formula C7H12BrNO2.
  • Suitable for muscarinic receptor pharmacology and neuropharmacology assays.
  • Supplied as a 10 mg solid for small-scale experimental use.