Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guvacoline hydrobrom 25mg | 17210-51-4 | 25 MG
SDP

Catalog No. 5000226200
Encompass_Preferred

Guvacoline hydrobromide is the hydrobromide salt of guvacoline, a pyridine alkaloid found in Areca triandra. It is used as a research reagent and functions as a weak full agonist of atrial and ileal muscarinic acetylcholine receptors (mAChR).

  • Acts as a weak full agonist of atrial and ileal muscarinic receptors.
  • White to off-white solid physical form.
  • Molecular formula C7H12BrNO2 and molecular weight 222.08.
  • Purity typically around 96.09% by HPLC.
  • Store sealed at 4°C and protect from moisture and light; in solution store at -80°C for long-term.
  • Supplied in small milligram pack sizes for laboratory research.

Catalog No. 50-002-26200 Supplier Medchemexpress LLC Supplier No. HY11649025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guvacoline hydrobromide is the hydrobromide salt of guvacoline, a pyridine alkaloid found in Areca triandra. It is used as a research reagent and functions as a weak full agonist of atrial and ileal muscarinic acetylcholine receptors (mAChR).

  • Acts as a weak full agonist of atrial and ileal muscarinic receptors.
  • White to off-white solid physical form.
  • Molecular formula C7H12BrNO2 and molecular weight 222.08.
  • Purity typically around 96.09% by HPLC.
  • Store sealed at 4°C and protect from moisture and light; in solution store at -80°C for long-term.
  • Supplied in small milligram pack sizes for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.