Learn More
Medchemexpress LLC Guvacoline hydrobrom 50mg | 17210-51-4 | 222.08 g·mol⁻¹ | C7H12BrNO2 | 50 MG
SDP
Catalog No. 5000308079
Encompass_Preferred
Guvacoline hydrobromide is a research-grade pyridine alkaloid that acts as a weak full agonist of atrial and ileal muscarinic acetylcholine receptors (mAChR). Supplied as a white to off-white solid with reported purity of 96.09%, it is used in receptor pharmacology and biochemical studies and is available in small laboratory pack sizes.
  • White to off-white solid suitable for laboratory use.
  • Reported purity of 96.09%.
  • Molecular weight 222.08 g·mol⁻¹.
  • Available in small pack sizes including 50 MG for bench-scale experiments.
  • Store sealed at 4°C away from moisture and light; in solvent keep at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-003-08079 Supplier Medchemexpress LLC Supplier No. HY11649050MG
Guvacoline hydrobromide is a research-grade pyridine alkaloid that acts as a weak full agonist of atrial and ileal muscarinic acetylcholine receptors (mAChR). Supplied as a white to off-white solid with reported purity of 96.09%, it is used in receptor pharmacology and biochemical studies and is available in small laboratory pack sizes.
  • White to off-white solid suitable for laboratory use.
  • Reported purity of 96.09%.
  • Molecular weight 222.08 g·mol⁻¹.
  • Available in small pack sizes including 50 MG for bench-scale experiments.
  • Store sealed at 4°C away from moisture and light; in solvent keep at -80°C (6 months) or -20°C (1 month).