Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guvacoline hydrobrom 5mg | 17210-51-4 | 222.08 g/mol | C7H12BrNO2 | 5 MG
SDP

Catalog No. 5000308080
Encompass_Preferred

Guvacoline hydrobromide is the hydrobromide salt of guvacoline, a pyridine alkaloid that acts as a weak full agonist of atrial and ileal muscarinic acetylcholine receptors (mAChR). It is supplied as a solid reagent intended for biochemical and pharmacological research applications.

  • Hydrobromide salt form suitable for laboratory handling
  • CAS number 17210-51-4
  • Molecular formula C7H12BrNO2; molecular weight 222.08 g/mol
  • Purity 96.09% as verified by certificate of analysis
  • Used as a pharmacological tool for muscarinic receptor studies

Catalog No. 50-003-08080 Supplier Medchemexpress LLC Supplier No. HY1164905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guvacoline hydrobromide is the hydrobromide salt of guvacoline, a pyridine alkaloid that acts as a weak full agonist of atrial and ileal muscarinic acetylcholine receptors (mAChR). It is supplied as a solid reagent intended for biochemical and pharmacological research applications.

  • Hydrobromide salt form suitable for laboratory handling
  • CAS number 17210-51-4
  • Molecular formula C7H12BrNO2; molecular weight 222.08 g/mol
  • Purity 96.09% as verified by certificate of analysis
  • Used as a pharmacological tool for muscarinic receptor studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.