Learn More
Medchemexpress LLC Guvacoline hydrochloride | 6197-39-3 | 1 ML
SDP
Catalog No. 5000419781
Encompass_Preferred
Guvacoline hydrochloride, a pyridine alkaloid isolated from Areca triandra, functions as a weak full agonist of atrial and ileal muscarinic receptors. This product is supplied as a 10 mM solution in DMSO, intended for research use only.
  • Pyridine alkaloid
  • Weak full agonist of atrial and ileal muscarinic receptors
  • Isolated from Areca triandra
  • Supplied as a 10 mM solution in DMSO
  • For research use only
  • Store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture and light.
Catalog No. 50-004-19781 Supplier Medchemexpress LLC Supplier No. HYN501610MM/1ML
Guvacoline hydrochloride, a pyridine alkaloid isolated from Areca triandra, functions as a weak full agonist of atrial and ileal muscarinic receptors. This product is supplied as a 10 mM solution in DMSO, intended for research use only.
  • Pyridine alkaloid
  • Weak full agonist of atrial and ileal muscarinic receptors
  • Isolated from Areca triandra
  • Supplied as a 10 mM solution in DMSO
  • For research use only
  • Store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture and light.