Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guvacoline hydrochloride | 6197-39-3 | 25 MG
SDP

Catalog No. 5000420518
Encompass_Preferred

Guvacoline hydrochloride is a pyridine alkaloid found in Areca triandra that acts as a weak full agonist of atrial and ileal muscarinic receptors.

  • Pyridine alkaloid
  • Found in Areca triandra
  • Acts as a weak full agonist of atrial and ileal muscarinic receptors
  • Purity: 98.0%
  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 177.63
  • Molecular formula: C7H12ClNO2
  • Storage: 4°C, sealed, away from moisture and light
  • Soluble in H2O: 100 mg/mL (needs ultrasonic)
  • Soluble in DMSO: 100 mg/mL (needs ultrasonic)
  • For research use only

Catalog No. 50-004-20518 Supplier Medchemexpress LLC Supplier No. 25MGHYN5016
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guvacoline hydrochloride is a pyridine alkaloid found in Areca triandra that acts as a weak full agonist of atrial and ileal muscarinic receptors.

  • Pyridine alkaloid
  • Found in Areca triandra
  • Acts as a weak full agonist of atrial and ileal muscarinic receptors
  • Purity: 98.0%
  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 177.63
  • Molecular formula: C7H12ClNO2
  • Storage: 4°C, sealed, away from moisture and light
  • Soluble in H2O: 100 mg/mL (needs ultrasonic)
  • Soluble in DMSO: 100 mg/mL (needs ultrasonic)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.