Learn More
TARGETMOL CHEMICALS INC GV-150013X 5MG
SDP
Catalog No. 502580605
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GV-150013X ((S)-1-(1-(adamantan-1-ylmethyl)-24-dioxo-5-phenyl-2345-tetrahydro-1H-benzo[b][14]diazepin-3-yl)-3-phenylurea) is a potent CCKB antiagonist pKi= 9.02. purity: 100%
Catalog No. 50-258-0605 Supplier TARGETMOL CHEMICALS INC Supplier No. T27503L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GV-150013X ((S)-1-(1-(adamantan-1-ylmethyl)-24-dioxo-5-phenyl-2345-tetrahydro-1H-benzo[b][14]diazepin-3-yl)-3-phenylurea) is a potent CCKB antiagonist pKi= 9.02. purity: 100%