Learn More
AdipoGen GV-58 (1 mg)
SDP
Supplier:  AdipoGen AGMRC0035M001
Encompass_Preferred
Chemical. Formula: C18H26N6OS. MW: 374.5. Selective N- and P/Q-type Ca2+-channel agonist. These Ca2+-channels regulate transmitter release in synapses. Potential lead compound for a variety of disorders that result in neuromuscular weakness.
Catalog No. 50-205-4254