Learn More
Gilson Company Inc ASTM E11 Calibration Test Sieve Verification
Catalog No. NC1244356
Encompass_Preferred
Calibration grade Test Sieve Verification ensures conformance to ASTM E11 specifications with a 99.73% standard deviation confidence level. Wire cloth openings are scanned automatically using an NIST traceable optical comparator to provide a statistical analysis and report.
Catalog No. NC1244356 Supplier Gilson Company Inc Supplier No. GV65
Calibration grade Test Sieve Verification ensures conformance to ASTM E11 specifications with a 99.73% standard deviation confidence level. Wire cloth openings are scanned automatically using an NIST traceable optical comparator to provide a statistical analysis and report.