Learn More
Enzo Life Sciences Gö 6976 (500µg). CAS: 136194-77-9
SDP
Supplier:  Enzo Life Sciences BMLEI2690500
Encompass_Preferred
Very potent protein kinase C (PKC) inhibitor with high selectivity for the PKC isotypes α and β. Inhibits PKCα (IC50=2.3nM) and PKCß1 (IC50=6.2nM) but has no effect on δ, ε or γ isotypes. Potent antagonist of HIV-1 induction. Alternative name: 12-(2-Cyanoethyl)-6,7,12,13-tetrahydro-13-methyl-5-oxo-5H-indolo[2,3-a]pyrrolo[3,4-c]carbazole. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0650