Learn More
INTACT GENOMICS INC GV3101 Agrobacterium ElectroCompetent Cells 18x50µl
Supplier:  INTACT GENOMICS INC 128236
Encompass
Intact Genomics GV3101 Agrobacterium ElectroCompetent Cells are optimized for the highest transformation efficiencies which is ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction.The GV3101 strain has a C58 chromosomal background with rifampicin resistance and the Ti plasmid pMP90 (pTiC58DT-DNA) with gentamicin resistance. The GV3101 Ti plasmid has the T-DNA region sequences deleted and transformation with a binary vector containing the missing T-region results in a functional T-DNA binary system that allows for transfer of genetic material into a host plant’s genome. Therefore, this system is often used for Agrobacterium-mediated transformation of several dicots such as Arabidopsis thaliana, tobacco, potatoes, and monocots like corn.Product Includes and Storage:GV3101 Agrobacterium ElectroCompetent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-82805