Learn More
Gold Biotechnology Inc GV3101 Chemically Competent Agrobacterium 5x50 μl
SDP
Supplier:  Gold Biotechnology Inc CC1055X50
Encompass_Preferred
GoldBio’s GV3101 Agrobacterium chemically competent cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our GV3101 strain harbors the C58 chromosomal backbone containing rifampicin resistance and the Ti plasmid pMP90 (pTiC58DT-DNA) harboring the gentamicin resistance. A functional T-DNA binary system can be built using our GV3101 strains as the T-DNA region has been deleted in the Ti plasmid and instead has a binary vector containing the missing T-region. The binary system makes possible to transfer genetic material into a host plant’s genome. Our system is often used for Agrobacterium-mediated transformation in mono and dicotyledonous species such as Arabidopsis thaliana, tobacco, potato, soybeans and corn.
Catalog No. NC2817839