Learn More
Gold Biotechnology Inc GV3101 pSoup Agro Electro
SDP
Catalog No. 5000562209
Encompass_Preferred
GoldBio s GV3101 Agrobacterium electrocompetent cells are a powerful tool for high efficiency transformation of plants The GV3101 strain is optimized for cDNA and gDNA library construction and possesses resistance to rifampicin gentamicin and tetracycline The T-DNA region of the GV3101 Ti plasmid has been deleted but restoration of the binary vector T-region can fully enable T-DNA binary system function
Catalog No. 50-005-62209 Supplier Gold Biotechnology Inc Supplier No. CC2175X50
GoldBio s GV3101 Agrobacterium electrocompetent cells are a powerful tool for high efficiency transformation of plants The GV3101 strain is optimized for cDNA and gDNA library construction and possesses resistance to rifampicin gentamicin and tetracycline The T-DNA region of the GV3101 Ti plasmid has been deleted but restoration of the binary vector T-region can fully enable T-DNA binary system function