Learn More
Gold Biotechnology Inc GV3101 pSoup Agro Electro
SDP
Catalog No. 5000562247
Encompass_Preferred
GoldBio s GV3101 Agrobacterium electrocompetent cells are a powerful tool for high efficiency transformation of plants The GV3101 strain is optimized for cDNA and gDNA library construction and possesses resistance to rifampicin gentamicin and tetracycline The T-DNA region of the GV3101 Ti plasmid has been deleted but restoration of the binary vector T-region can fully enable T-DNA binary system function
Catalog No. 50-005-62247 Supplier Gold Biotechnology Inc Supplier No. CC21710X50
GoldBio s GV3101 Agrobacterium electrocompetent cells are a powerful tool for high efficiency transformation of plants The GV3101 strain is optimized for cDNA and gDNA library construction and possesses resistance to rifampicin gentamicin and tetracycline The T-DNA region of the GV3101 Ti plasmid has been deleted but restoration of the binary vector T-region can fully enable T-DNA binary system function